Potassium hydroxide 1 mol/L (1 N), AVS TITRINORM (max. 0.00005% Ca)
31309.290
:
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
- Titer (20°C; real value 0.2 % accuracy):0,998 - 1,002 mol/l
- Ca (Calcium):≤ 0,00005 %
- NIST traceable:Confirmed
: Potash caustic, Caustic potash
:
:56.11 g/mol
:1814
:GHS05
:Ambient
:1310-58-3
:KOH
:MFCD00003553
:1320
:360
:8 II
:12,07806