Order Entry
Belgium
ContactUsLinkComponent
DNA isolation, tissue and cells genomicPrep mini spin kit, illustra™
DNA isolation, tissue and cells genomicPrep mini spin kit, illustra™
Supplier:  Cytiva
Certificates
undefined
DNA isolation, tissue and cells genomicPrep mini spin kit, illustra™
Supplier:  Cytiva

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    illustra™ tissue and cells genomicPrep Mini Spin Kit

Specifications

Frequently Bought Together