Orders LinkContactUsLinkComponent
epT.I.P.S.® Racks - Pipette Tips
epT.I.P.S.® Racks - Pipette Tips
   Sustainable Options Available
Supplier:   EPPENDORF
epT.I.P.S.® Racks - Pipette Tips
Supplier:  EPPENDORF
   Sustainable Options Available
Certificates
epT.I.P.S.® Racks - Pipette Tips
14 Options Available Below
Product Details & Documents
Eppendorf epT.I.P.S.® Racks feature high-precision pipette tips in 'Eppendorf Quality', 'PCR clean', 'Biopur®', and 'Sterile' purity grades. All tips are supplied in disposible racks designed for optimal organisation and user convenience in demanding laboratory environments.
  • Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
  • Modern slim rack design with reclosable lid
  • Improved sustainability through 35% lower polypropylene consumption per rack
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally integrated pipetting system') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.
How to recognise an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.
Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0,5 to 20 µl, 50 to 1250 µl, 0,2 to 5 ml, and 0,5 to 10 ml.
Certifications: Lot-specific certficates available for the purity grades PCR clean, Sterile, and Biopur® can be downloaded from the Eppendorf website.
Specifications for Product Family
Specifications
  • Catalog No:
  • 613-6933
  • 613-6934
  • 613-6935
  • 613-6936
  • 613-6751
  • 613-6752
  • 613-6753
  • 613-6754
  • 613-6755
  • 613-6756
  • 613-6757
  • 613-6758
  • 613-6759
  • 613-6760
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Specifications
Product Family Options
Eppendorf quality (non-sterile)
Product Information
  • Volume
    Length
    Colour code
    Packed
    Availability
    Price
  • 100 - 5000 µl
    120 mm
    Violet
    120 tips (5 racks×24 tips)
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 46,80 €
    46,80 €
    1 * 120 ST
       46,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-6933
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 200 - 5000 µl
    175 mm
    Violet
    120 tips (5 racks×24 tips)
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 57,30 €
    57,30 €
    1 * 120 ST
       57,30 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-6934
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 500 - 10000 µl
    165 mm
    Turquoise
    120 tips (5 racks×24 tips)
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 70,20 €
    70,20 €
    1 * 120 ST
       70,20 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-6935
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
PCR clean (non-sterile)
Product Information
  • Volume
    Length
    Colour code
    Packed
    Availability
    Price
  • 100 - 5000 µl
    120 mm
    Violet
    120 tips (5 racks×24 tips)
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 55,40 €
    55,40 €
    1 * 120 ST
       55,40 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-6936
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
Biopur® (sterile)
Product Information
  • Volume
    Length
    Colour code
    Packed
    Availability
    Price
  • 0,1 - 20 µl
    40 mm
    Grey
    480 tips (5 racks×96 tips)
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 67,70 €
    67,70 €
    1 * 480 ST
       67,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-6751
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 2 - 200 µl
    53 mm
    Yellow
    480 tips (5 racks×96 tips)
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 63,00 €
    63,00 €
    1 * 480 ST
       63,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-6752
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 20 - 300 µl
    55 mm
    Orange
    480 tips (5 racks×96 tips)
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 63,00 €
    63,00 €
    1 * 480 ST
       63,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-6753
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 50 - 1000 µl
    71 mm
    Blue
    480 tips (5 racks×96 tips)
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 63,00 €
    63,00 €
    1 * 480 ST
       63,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-6754
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 50 - 1250 µl
    76 mm
    Green
    480 tips (5 racks×96 tips)
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 63,00 €
    63,00 €
    1 * 480 ST
       63,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-6755
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 50 - 1250 µl
    103 mm
    Dark green
    480 tips (5 racks×96 tips)
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 63,00 €
    63,00 €
    1 * 480 ST
       63,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-6756
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 100 - 2500 µl (0,1 - 2,5 ml)
    115 mm
    Red
    240 tips (5 racks×48 tips)
    Log in for availability
    Log in for availability
    1 * 240 ST
    1 * 240 ST - 56,30 €
    56,30 €
    1 * 240 ST
       56,30 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-6757
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 100 - 5000 µl
    120 mm
    Violet
    120 tips (5 racks×24 tips)
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 62,70 €
    62,70 €
    1 * 120 ST
       62,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-6758
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 200 - 5000 µl
    175 mm
    Violet
    120 tips (5 racks×24 tips)
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 72,80 €
    72,80 €
    1 * 120 ST
       72,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-6759
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 500 - 10000 µl
    165 mm
    Turquoise
    120 tips (5 racks×24 tips)
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 75,10 €
    75,10 €
    1 * 120 ST
       75,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-6760
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact