Order Entry
Belgium
ContactUsLinkComponent
Chromate standard solution, traceable to SRM of NIST, Certipur®, Supelco®
Chromate standard solution, traceable to SRM of NIST, Certipur®, Supelco®
Catalog # 1.19780.0500
Supplier:  Merck
CAS Number:  
Chromate standard solution, traceable to SRM of NIST, Certipur®, Supelco®
Catalog # 1.19780.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Chromate standard solution
  • M-Clarity quality level:
    MQ100
  • Contents:
    K₂CrO₄ in H₂O

Specifications

About this item