CPC systems
About this item
Gilson’s reusable, silica-free centrifugal partition chromatography (CPC) columns offer high injection capacities of milligram to multi-kilograms resulting in 95% recoveries with 99% purity or better.
- Increased savings
- Improved purification efficiency
- Numerous column options
CPC columns use approximately five times less solvent than more traditional columns, saving you money on solvent and disposal costs. Likewise, the reusable columns eliminates column replacement and silica recycling costs. CPC columns offer high sample injection capacities from milligrams to kilograms. With these injection rates, achieve 95% recoveries with greater than 99% purity because there is no irreversible absorption or denaturation of fragile molecules as in other methods. CPC columns tackle tough separations of complex samples in a single step without prior purification, making it an ideal choice for natural product isolation, protein purification, and more. Choose from a variety of injection capacities and flow rates to optimise your purification with the CPC. All CPC columns can be connected to either a PLC purification system or a Gilson modular HPLC system in place of a traditional column.
3 Options Available Below
Product Details & Documents
Gilson’s reusable, silica-free centrifugal partition chromatography (CPC) columns offer high injection capacities of milligram to multi-kilograms resulting in 95% recoveries with 99% purity or better.
- Increased savings
- Improved purification efficiency
- Numerous column options
CPC columns use approximately five times less solvent than more traditional columns, saving you money on solvent and disposal costs. Likewise, the reusable columns eliminates column replacement and silica recycling costs. CPC columns offer high sample injection capacities from milligrams to kilograms. With these injection rates, achieve 95% recoveries with greater than 99% purity because there is no irreversible absorption or denaturation of fragile molecules as in other methods. CPC columns tackle tough separations of complex samples in a single step without prior purification, making it an ideal choice for natural product isolation, protein purification, and more. Choose from a variety of injection capacities and flow rates to optimise your purification with the CPC. All CPC columns can be connected to either a PLC purification system or a Gilson modular HPLC system in place of a traditional column.
Product Family Options
Product Information
- CapacityFlow rateAvailabilityPrice
- GILI21141007CPC 250, 230 V, 100 bar (1450 psi) max. pressureCPC 250, 230 V, 100 bar (1450 psi) max. pressureUp to 6 g (injection cap.)15 ml/min. (max. elution)Log in for availability1 * 1 ST86 666,67 €
- GILI21141008CPC 1000, 230 V, 80 bar (1160 psi) max. pressureCPC 1000, 230 V, 80 bar (1160 psi) max. pressureUp to 30 g (injection cap.)50 ml/min. (max. elution)Log in for availability1 * 1 ST118 666,67 €
- GILI21141010CPC 1000 PRO, 230 V, 80 bar (1160 psi) max. pressureCPC 1000 PRO, 230 V, 80 bar (1160 psi) max. pressureUp to 100 g (injection cap.)350 ml/min. (max. elution)Log in for availability1 * 1 ST147 801,33 €
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.