Orders LinkContactUsLinkComponent
PLC purification systems
PLC purification systems
Fournisseur:   GILSON LCMS
PLC purification systems
Fournisseur:  GILSON LCMS
Certificats
PLC purification systems
4 Options disponibles ci-dessous
Product Details & Documents
PLC 2050/2250/2500 purification systems are designed for preparative liquid chromatography (PLC) to aspirate mobile phase solvents, form gradients, inject liquid samples, and provide solutions for a preparative column for high performance liquid chromatography (HPLC), centrifugal partition chromatography (CPC), or counter-current chromatography (CCC) applications.
  • PLC 2050: Flow rates up to 50 ml/min with pressure up to 300 bar
  • PLC 2250: Flow rates up to 250 ml/min with pressure up to 230 bar
  • PLC 2500: Flow rates up to 500 ml/min with pressure up to 110 bar
  • Intuitive touchscreen interface for easy method writing and modyfying of parameters
  • Autosampler available for automated sample injections for increased throughput
  • Costs: 'All in one' system
  • Safety: Compact footprint that fits into a fume hood
With the capability to purify compounds by preparative HPLC and flash chromatography on the same instrument, Gilson's PLC 2050/2250/2500 purification systems simplify and streamline compound purification. These compact systems feature UV/VIS detection with an optional upgrade to a diode array detector (DAD) or evaporative light scattering detection (ELSD), an integrated pumping system, fraction collector, touchscreen control, and Gilson glider software. These customisable instruments are available in simple configurations for basic applications, but with a wide range of upgrade options to meet your lab’s demands.
Electrical: 110 to 120 / 220 to 240 V
Product Family Options
Informations sur le produit
  • Disponibilité
    Prix
  • GILI21140001
    PLC 2050 purification system, 1 - 50 ml/min. flow rate, 300 bar max. pressure
    PLC 2050 purification system, 1 - 50 ml/min. flow rate, 300 bar max. pressure
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 47 760,90 €
    47 760,90 €
    1 * 1 ST
       47 760,90 €
  • GILI21140002
    PLC 2250 purification system, 5 - 250 ml/min. flow rate, 230 bar max. pressure
    PLC 2250 purification system, 5 - 250 ml/min. flow rate, 230 bar max. pressure
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 47 760,90 €
    47 760,90 €
    1 * 1 ST
       47 760,90 €
  • GILI21140004
    PLC 2050 with autosampler, 1 - 50 ml/min. flow rate, 300 bar max. pressure
    PLC 2050 with autosampler, 1 - 50 ml/min. flow rate, 300 bar max. pressure
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 78 862,30 €
    78 862,30 €
    1 * 1 ST
       78 862,30 €
  • GILI21140005
    PLC 2250 with autosampler, 5 - 250 ml/min., 230 bar max. pressure
    PLC 2250 with autosampler, 5 - 250 ml/min., 230 bar max. pressure
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 78 862,30 €
    78 862,30 €
    1 * 1 ST
       78 862,30 €
Accessories
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Rack set of 3 racks, 25×150 mm tubes
    Rack set of 3 racks, 25×150 mm tubes
    GILI21040023
    Connexion Pour la disponibilité
    1 * 3 ST - 1 497,10 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Rack set of 3 racks, 28×150 mm tubes
    Rack set of 3 racks, 28×150 mm tubes
    GILI21040024
    Connexion Pour la disponibilité
    1 * 3 ST - 1 497,10 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Rack set of 3 racks, 29,5×200 mm tubes
    Rack set of 3 racks, 29,5×200 mm tubes
    GILI21040025
    Connexion Pour la disponibilité
    1 * 3 ST - 1 497,10 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Rack set of 3 racks, 13×73 mm hemo tubes
    Rack set of 3 racks, 13×73 mm hemo tubes
    GILI21040026
    Connexion Pour la disponibilité
    1 * 3 ST - 1 497,10 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Rack funnel, 16 outlets ¹/₄" Ext.Ø
    Rack funnel, 16 outlets ¹/₄" Ext.Ø
    GILI21040027
    Connexion Pour la disponibilité
    1 * 3 ST - 2 795,10 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Sample loop, 40 ml auto, ¹/₈" Ext.Ø
    Sample loop, 40 ml auto, ¹/₈" Ext.Ø
    GILI21040036
    Connexion Pour la disponibilité
    1 * 1 ST - 837,10 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Sample loop, 50 ml auto, ¹/₈" Ext.Ø
    Sample loop, 50 ml auto, ¹/₈" Ext.Ø
    GILI21040037
    Connexion Pour la disponibilité
    1 * 1 ST - 1 006,50 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Manual injection valve with tubing kit (PLC 2050)
    Manual injection valve with tubing kit (PLC 2050)
    GILI21040181
    Connexion Pour la disponibilité
    1 * 1 ST - 3 106,40 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Manual injection valve with tubing kit (PLC 2250/2500)
    Manual injection valve with tubing kit (PLC 2250/2500)
    GILI21040182
    Connexion Pour la disponibilité
    1 * 1 ST - 3 106,40 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Injection pump Ext (50 ml/min, 300 Bar)
    Injection pump Ext (50 ml/min, 300 Bar)
    GILI21040184
    Connexion Pour la disponibilité
    1 * 1 ST - 16 397,70 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Rack set of 3 racks, 12×65 mm tubes
    Rack set of 3 racks, 12×65 mm tubes
    GILI21040191
    Connexion Pour la disponibilité
    1 * 3 ST - 1 497,10 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Rack set of 3 racks, 18×150/180 mm tubes
    Rack set of 3 racks, 18×150/180 mm tubes
    GILI21040019
    Connexion Pour la disponibilité
    1 * 3 ST - 1 075,80 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Rack set of 3 racks, 13×100 mm tubes
    Rack set of 3 racks, 13×100 mm tubes
    GILI21040020
    Connexion Pour la disponibilité
    1 * 3 ST - 1 497,10 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Rack set of 3 racks, 16×150 mm tubes
    Rack set of 3 racks, 16×150 mm tubes
    GILI21040021
    Connexion Pour la disponibilité
    1 * 3 ST - 1 497,10 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Rack set of 3 racks, 21×150 mm tubes
    Rack set of 3 racks, 21×150 mm tubes
    GILI21040022
    Connexion Pour la disponibilité
    1 * 3 ST - 1 497,10 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • 3 Rack overlays for 18 mm tubes
    3 Rack overlays for 18 mm tubes
    GILI21040028
    Connexion Pour la disponibilité
    1 * 3 ST - 168,30 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • 3 Rack overlays 13 mm tubes
    3 Rack overlays 13 mm tubes
    GILI21040029
    Connexion Pour la disponibilité
    1 * 3 ST - 168,30 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Sample loop, 1 ml auto, ¹/₈" Ext.Ø
    Sample loop, 1 ml auto, ¹/₈" Ext.Ø
    GILI21040030
    Connexion Pour la disponibilité
    1 * 1 ST - 146,30 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Sample loop, 2 ml auto, ¹/₈" Ext.Ø
    Sample loop, 2 ml auto, ¹/₈" Ext.Ø
    GILI21040031
    Connexion Pour la disponibilité
    1 * 1 ST - 143,73 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Sample loop, 5 ml auto, ¹/₈" Ext.Ø
    Sample loop, 5 ml auto, ¹/₈" Ext.Ø
    GILI21040032
    Connexion Pour la disponibilité
    1 * 1 ST - 262,90 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Sample loop, 10 ml auto, ¹/₈" Ext.Ø
    Sample loop, 10 ml auto, ¹/₈" Ext.Ø
    GILI21040033
    Connexion Pour la disponibilité
    1 * 1 ST - 346,50 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Sample loop, 20 ml auto, ¹/₈" Ext.Ø
    Sample loop, 20 ml auto, ¹/₈" Ext.Ø
    GILI21040034
    Connexion Pour la disponibilité
    1 * 1 ST - 570,90 €
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Sample loop, 30 ml auto, ¹/₈" Ext.Ø
    Sample loop, 30 ml auto, ¹/₈" Ext.Ø
    GILI21040035
    Connexion Pour la disponibilité
    1 * 1 ST - 667,70 €