Order Entry
Belgium
ContactUsLinkComponent
Super rapid readout biological indicator 1492V, Attest™
Super rapid readout biological indicator 1492V, Attest™
  MMME1492V
Super rapid readout biological indicator 1492V, Attest™
  MMME1492V

 

  • Description:
    3M™ Attest™ Super rapid readout biological indicator, model 1492V
  • Pk:
    50

 

 

3M™ Attest™ Super rapid readout biological indicators and challenge packs for steam, super rapid 1-hour results.

  • Provides biological indicator results in 1 hour
  • Helps deliver BI results in about the same time as it takes to cool a load
  • Incubation period: 24 minutes

Offers three times faster results than Attest Rapid BIs for 132,2 and 135 °C (270 and 275 °F) vacuum-assisted steam sterilisation cycles.