ContactUsLinkComponent
Earmuffs, WS™ LiteCom Pro III Headset, Peltor™
Earmuffs, WS™ LiteCom Pro III Headset, Peltor™
Catalog # 111-2496
Supplier:  3M
undefined
Earmuffs, WS™ LiteCom Pro III Headset, Peltor™
Catalog # 111-2496
Supplier:  3M
Specifications
  • Colour:
    Black/blue
  • Description:
    Peltor™ WS™ LiteCom Pro III Headset, 403-470 MHz, analog/digital, EU version, headband
  • Pk:
    1
Specifications
About this item
The 3M™ Peltor™ WS™ LiteCom Pro III Headset features an integrated, programmable analog/digital two-way radio, Bluetooth® Multipoint connectivity, noise-cancelling microphone, and level-dependent function for ambient listening. The headset not only helps protect against hazardous noise, but also makes it easy to communicate in noisy environments and maintain situational awareness.
  • Programmable analog/digital DMR two-way radio communication
  • Bluetooth® multipoint functionality
  • Water tight microphone (IP 68)
  • Level-dependent function for ambient listening
  • Voice menu with multiple language selection
  • Push-to-Listen function
  • Rechargeable battery, Li-Ion
  • Voice operated transmission (VOX)
The Peltor™ WS™ LiteCom Pro III Headset offers wireless communication in noisy environments. Suitable for work in industrial and construction applications, where hands-free communication and hearing protection are needed. Additional features include busy channel indication, an external connection jack, auto power off function, and battery status indication.
Certifications: Conforms with the provisions set out in the PPE directive 89/686/EEC and RED directive 2014/53/EU.