Orders LinkContactUsLinkComponent
Spinner flasks with angled sidearms, Corning®
Spinner flasks with angled sidearms, Corning®
Supplier:   Corning
Spinner flasks with angled sidearms, Corning®
Supplier:  Corning
Certificates
Spinner flasks with angled sidearms, Corning®
Spinner flasks with angled sidearms, Corning®
14 Options Available Below
Product Details & Documents
Borosilicate glass with side arms, non sterile.
  • Baffles enhance aeration and agitation of flask contents
  • Impeller design ensures optimal stirring
  • Complete with impeller and caps
  • Side arm design permit easy access of 25 and 50 ml pipettes
Documents
Product Family Options
Product Information
  • Capacity
    Neck
    No. of side arms
    Availability
    Price
  • 125 ml
    Centre neck 70 mm, side arm neck 32 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 774,00 €
    774,00 €
    1 * 1 ST
       774,00 €
  • 15000 ml
    Centre neck 100 mm, side arm neck 45 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 440,00 €
    2 440,00 €
    1 * 1 ST
       2 440,00 €
  • 1000 ml
    Centre neck 100 mm, side arm neck 45 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 220,00 €
    1 220,00 €
    1 * 1 ST
       1 220,00 €
  • 250 ml
    Centre neck 70 mm, side arm neck 32 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 803,00 €
    803,00 €
    1 * 1 ST
       803,00 €
  • 36000 ml
    Centre neck 100 mm, side arm neck 45 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 570,00 €
    3 570,00 €
    1 * 1 ST
       3 570,00 €
  • 3000 ml
    Centre neck 100 mm, side arm neck 45 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 390,00 €
    1 390,00 €
    1 * 1 ST
       1 390,00 €
  • 500 ml
    Centre neck 100 mm, side arm neck 45 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 848,00 €
    848,00 €
    1 * 1 ST
       848,00 €
  • 6000 ml
    Centre neck 100 mm, side arm neck 45 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 630,00 €
    1 630,00 €
    1 * 1 ST
       1 630,00 €
  • 8000 ml
    Centre neck 100 mm, side arm neck 45 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 890,00 €
    1 890,00 €
    1 * 1 ST
       1 890,00 €
  • 15000 ml
    Centre neck 120 mm, side arm neck 45 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 460,00 €
    2 460,00 €
    1 * 1 ST
       2 460,00 €
  • 36000 ml
    Centre neck 120 mm, side arm neck 45 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 680,00 €
    3 680,00 €
    1 * 1 ST
       3 680,00 €
  • 3000 ml
    Centre neck 120 mm, side arm neck 45 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 450,00 €
    1 450,00 €
    1 * 1 ST
       1 450,00 €
  • 6000 ml
    Centre neck 120 mm, side arm neck 45 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 740,00 €
    1 740,00 €
    1 * 1 ST
       1 740,00 €
  • 8000 ml
    Centre neck 120 mm, side arm neck 45 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 960,00 €
    1 960,00 €
    1 * 1 ST
       1 960,00 €