Orders LinkContactUsLinkComponent
Spinner flasks, disposable, Corning®
Spinner flasks, disposable, Corning®
Supplier:   Corning
Spinner flasks, disposable, Corning®
Supplier:  Corning
Certificates
Spinner flasks, disposable, Corning®
Spinner flasks, disposable, Corning®
7 Options Available Below
Product Details & Documents
The Corning® disposable spinner flask system comes ready-to-use with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly.
  • Paddle size and height is optimised for each vessel size
  • Integrated magnet provides smooth, even rotation at required speeds on slow-speed stirrers
  • Heat build-up in the vessel is reduced by means of a specially designed flange that raised the vessel off the stir-plate surface for the 125 and 500 ml flasks only
Molded from virgin polystyrene and gamma-irradiated, each spinner flask system assures a clean and sterile unit that is nonpyrogenic. No more concerns with detergent residues or contamination.
Made of USP Class VI and ISO 10993-18 polystyrene, the vessel is comparable to conventional glass spinner flasks for growth of suspension cell lines and any attachment-dependent cultures using microcarrier beads.
Product Family Options
Product Information
  • Capacity
    Neck
    No. of side arms
    Recommended working volume
    Ø×H
    Availability
    Price
  • 734-4049
    Flasks with two solid caps on the side arms
    Flasks with two solid caps on the side arms
    125 ml
    centre neck 70 mm Ø, side arm neck thread GL 25
    2
    30-75 ml
    63,5×145 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 1 350,00 €
    1 350,00 €
    1 * 12 ST
       1 350,00 €
  • 734-4050
    Flasks with two solid caps on the side arms
    Flasks with two solid caps on the side arms
    500 ml
    centre neck 100 mm Ø, side arm neck thread GL 45
    2
    100-300 ml
    87,3× 203,2 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 1 170,00 €
    1 170,00 €
    1 * 12 ST
       1 170,00 €
  • 392-0309
    Flasks with two vented caps on the side arms
    Flasks with two vented caps on the side arms
    500 ml
    centre neck 100 mm Ø, side arm neck thread GL 45
    2
    100-300 ml
    87,3× 203,2 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 1 350,00 €
    1 350,00 €
    1 * 12 ST
       1 350,00 €
  • 734-4097
    Flasks with two solid caps on the side arms
    Flasks with two solid caps on the side arms
    1000 ml
    side arm necks thread GL 45
    2
    300-1000 ml
    137×245 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 1 390,00 €
    1 390,00 €
    1 * 6 ST
       1 390,00 €
  • 734-2289
    Flasks with two vented caps on the side arms
    Flasks with two vented caps on the side arms
    1000 ml
    side arm necks thread GL 45
    2
    300-1000 ml
    137×245 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 1 390,00 €
    1 390,00 €
    1 * 6 ST
       1 390,00 €
  • 734-4099
    Flasks with two solid caps on the side arms
    Flasks with two solid caps on the side arms
    3000 ml
    side arm necks thread GL 45
    2
    1000-3000 ml
    188×259 mm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 1 130,00 €
    1 130,00 €
    1 * 4 ST
       1 130,00 €
  • 734-2290
    Flasks with two vented caps on the side arms
    Flasks with two vented caps on the side arms
    3000 ml
    side arm necks thread GL 45
    2
    1000-3000 ml
    188×259 mm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 1 130,00 €
    1 130,00 €
    1 * 4 ST
       1 130,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Modified impeller assembly for magnetically-driven 36 L Corning® Proculture® spinner flask, reusable
    Modified impeller assembly for magnetically-driven 36 L Corning® Proculture® spinner flask, reusable
    CORN401901
    Log in for availability
    1 * 1 ST - 1 340,00 €
Product Information
  • Catalog #
    Thread
    Availability
    Price
  • Vent cap, 0,2 μm vent, for 500, 1000 and 3000 ml spinner flasks
    Vent cap, 0,2 μm vent, for 500, 1000 and 3000 ml spinner flasks
    734-4102
    GL45
    Log in for availability
    1 * 4 ST - 44,90 €
Product Information
  • Catalog #
    Thread
    Availability
    Price
  • Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 500 ml spinner flask
    Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 500 ml spinner flask
    734-4101
    GL45
    Log in for availability
    1 * 2 ST - 449,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Flat cap, PE, disposable, orange, for 500, 1000 and 3000 ml spinner flasks
    Flat cap, PE, disposable, orange, for 500, 1000 and 3000 ml spinner flasks
    CORN3568
    Log in for availability
    1 * 4 ST - 41,20 €
Product Information
  • Catalog #
    Thread
    Availability
    Price
  • Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 3000 ml spinner flask
    Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 3000 ml spinner flask
    734-4100
    GL45
    Log in for availability
    1 * 2 ST - 449,00 €
Product Information
  • Catalog #
    Thread
    Availability
    Price
  • Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 1000 ml spinner flask
    Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 1000 ml spinner flask
    734-4098
    GL45
    Log in for availability
    1 * 2 ST - 449,00 €