Welding boot cover, SW32
Catalog # PRTWSW32TAR
Supplier: Portwest
Specifications
- Garment application:Welding
- Gender:Unisex
- Risk area:Heat
- Colour:Tan
- Fabric:Split cowhide leather
- Pk:1 PAIR
Specifications
About this item
Boot cover made of split cowhide leather. The strong material protects the feet and lower legs when welding and is designed to last in the toughest environments.
- Class 2 welding protection
- Designed with a comfort fit
- Designed to be worn in a tough work environment
Certifications: EN ISO 11611 Class 2 A1