ASPEC® 241 system
Product Details & Documents
This compact ASPEC® system combines positive pressure solid phase extraction (SPE) with small footprint and precise liquid handling for reliable automated SPE applications.
- Positive pressure for reproducible extractions
- Small footprint for minimised performance
- Intuitive error handling for trusted results
- Powerful software control for optimised productivity
- Versatile set-up for trace analysis
With its minimised footprint, it is the ideal choice for basic SPE automation on 1, 3 or 6 ml cartridges for labs where bench space or throughput is limited. The syringe pump controls the accuracy of sample and solvent volumes. Combined with ASPEC SPE cartridges, the system generates reproducible sample clean-up for clinical, forensic, doping, food and environmental research labs.
Positive pressure for reproducible extractions:
Positive pressure extraction with unique sealing cap design increases reproducibility from sample to sample. The syringe pump delivers accurate volume dispensing and full control of sorbent drying.
Smart design for maximised performance:
The mobile racks accommodate 1, 3, and 6 ml SPE polypropylene and glass cartridges and enable multiple fraction elutions per sample. The syringe pump manages volumes from 1 to 10 ml with high accuracy and precision.
Intuitive error handling for trusted results:
The syringe pump's pressure-sensing module provides error handling for high cartridge pressure or clogged liquid lines, and monitors pressure on each fluid path individually. Optional jet wash rinse stations minimise carry-over and cross contamination.
Versatile software control for optimised productivity:
TRILUTION® Software enables liquid handling and SPE applications on the same ASPEC system. Optimise throughput and efficiency by processing samples in either batch or sequential mode. Multiple fraction elution allows you to automate and optimise method development.
Specifications
- Description:ASPEC 241 system, single syringe dilutor, standard volume
Complementary Products
Product Information
- Catalog #DescriptionAvailabilityPrice
Accessories
Product Information
- Catalog #AvailabilityPrice
- GX-241 ASPEC kit, 1 ml
Product Information
- Catalog #AvailabilityPrice
- GX-241 ASPEC kit, 3 ml
Product Information
- Catalog #AvailabilityPrice
- GX-241 ASPEC kit, 6 ml
Product Information
- Catalog #AvailabilityPrice
- Probe, 221×2,0×0,8 mm, septum piercing, beveled, grooved, 1,5 mm tipProbe, 221×2,0×0,8 mm, septum piercing, beveled, grooved, 1,5 mm tipGILI27067382Log in for availability1 * 1 ST - 258,67 €
Product Information
- Catalog #AvailabilityPrice
- Rack, code 330, holds 60 12×32 mm vials (2 ml)Rack, code 330, holds 60 12×32 mm vials (2 ml)GILI260440031Log in for availability1 * 1 ST - 285,60 €
Product Information
- Catalog #AvailabilityPrice
- Rack, code 334, holds 14 40 ml scintilation vialsRack, code 334, holds 14 40 ml scintilation vialsGILI260440081Log in for availability1 * 1 ST - 438,90 €
Product Information
- Catalog #AvailabilityPrice
- Rack, code 335, holds 48 15×45 mm wisp vials (4 ml)Rack, code 335, holds 48 15×45 mm wisp vials (4 ml)GILI260440083Log in for availability1 * 1 ST - 445,50 €
Product Information
- Catalog #AvailabilityPrice
- Rack, code 338, holds 64 12×32 mm vials (2 ml)Rack, code 338, holds 64 12×32 mm vials (2 ml)GILI260440106Log in for availability1 * 1 ST - 445,50 €
Product Information
- Catalog #AvailabilityPrice
- Rack, code 341, holds 108 10×75 mm tubes
Product Information
- Catalog #AvailabilityPrice
- Rack, code 343, holds 80 13×100 mm tubes
Product Information
- Catalog #AvailabilityPrice
- Rack, code 345, holds 44 16×150 mm tubes
Product Information
- Catalog #AvailabilityPrice
- Rack, code 346, holds 44 16×100 mm tubes
Product Information
- Catalog #AvailabilityPrice
- Solvent reservoir, 100 ml, ASPEC 241
Product Information
- Catalog #AvailabilityPrice
- Pressure regulator assay, 0 - 30 psi, ASPECPressure regulator assay, 0 - 30 psi, ASPECGILI25051376Log in for availability1 * 1 ST - 611,60 €
Product Information
- Catalog #AvailabilityPrice
- Syringe, 10 ml, 215/235
Product Information
- Catalog #AvailabilityPrice
- Tubes glass, 12×75 mm, 6 ml, 250/pack
Product Information
- Catalog #AvailabilityPrice
- Tubes glass, 15×85 mm, 10 ml, 250/pack
Product Information
- Catalog #AvailabilityPrice
- Plumbing package, air-gas, GX274 ASPEC
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



