Flame-retardant jackets, BizFlame™ Rain, S770 multiprotection
Supplier: Portwest
Certificates
About this item
This jacket is constructed from 98% polyester and 2% anti-static carbon fibre. The fabric has flame-resistant and anti-static properties and is therefore used for wide range of products suitable for all climates and end uses.
- Flame resistant treated waterproof fabric prevents water penetration
- Breathable fabric to draw moisture away from the body keeping the wearer cool, dry and comfortable
- Detachable lining for added versatility in all weathers conditions
- Taped seams to provide additional protection
- Premium sew on flame resistant reflective tape
- Two-way zip for quick and easy access
- Five pockets for ample storage with concealed phone pocket
These waterproof, breathable parka offers protection in multi-hazard environments. The detachable, FR fleece liner offers additional warmth in extreme weather conditions.
Certifications: EN ISO 14116 Index 3, EN 1149 -5 , EN 343:2019 Class 3:3 X, EN 13034 Type 6
Caution: Use non-biological powder. Do not use softener.
Specifications
- Catalog No:
- PRTWS770NARXL
- PRTWS770NARL
- PRTWS770NARXXXL
- PRTWS770NARM
- PRTWS770NARS
- PRTWS770NARXXL
- Model:
- S770
- S770
- S770
- S770
- S770
- S770
- Garment application:
- Fire protection, Outdoor
- Fire protection, Outdoor
- Fire protection, Outdoor
- Fire protection, Outdoor
- Fire protection, Outdoor
- Fire protection, Outdoor
- Risk area:
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- 98% polyester/2% anti-static carbon
- 98% polyester/2% anti-static carbon
- 98% polyester/2% anti-static carbon
- 98% polyester/2% anti-static carbon
- 98% polyester/2% anti-static carbon
- 98% polyester/2% anti-static carbon
- Weight:
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
Specifications
Frequently Bought Together