ContactUsLinkComponent
Fall protection harnesses, 2 point with chest loops
Fall protection harnesses, 2 point with chest loops
Catalog # PRTWFP12RER
Supplier:  Portwest
undefined
Fall protection harnesses, 2 point with chest loops
Catalog # PRTWFP12RER
Supplier:  Portwest
Specifications
  • Attachment Points:
    2-point
  • Description:
    Harness with chest loop
  • Pk:
    1
  • Weight:
    1,0 kg
Specifications
About this item
Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collision and restricting users from fall hazard areas.
  • Made from polyester webbing, alloy steel, heat treated
  • Light weight
  • Dorsal D-ring
  • Chest and leg adjustment points
Features two chest loops and an adjustable quick release chest strap.
Certifications: EN 361