Browguard with mesh visor, PW93
Galvanised carbon steel, PP browguard with mesh visor, PW93, 70×47 mm
Catalog # PRTWPW93BKR
Supplier: Portwest
Jump to:
Product Details & Documents
Mesh visor with PP browguard, suitable for protection against large highspeed particles. The ratchet adjustment system is designed for easy use.
- Wheel ratchet size adjustment for easy fitting
- Soft foam sweat band included
- Can be adapted to be used over prescription glasses
- Individually packed for vending machines
Materials: galvanised carbon steel, polypropylene.
Certifications: EN 1731 (F); EN 166 (B); CE
Specifications
- Description:Galvanised carbon steel, PP browguard with mesh visor
- L×W:70×47 mm
- Model:PW93