Orders LinkContactUsLinkComponent
Goggles, indirect vent
Goggles, indirect vent
UV protection, Clear PC, Transparent
Catalog # PRTWPW21CLR
Supplier:   Portwest
Goggles, indirect vent
Catalog # PRTWPW21CLR
Supplier:  Portwest
Goggles, indirect vent
Goggles, indirect vent
UV protection, Clear PC, Transparent
Price
3,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
Designed to be worn in a tough work environment, flexible PVC frame for ultra comfort, transparent colour for better side visibility.
  • 99% UV protection
  • Back elastic for ease of movement
  • Ultra lightweight for long uses
  • Individually packed for vending machines
  • Indirect ventilation
 : EN 166 (1 B 3); ANSI/ISEA Z87.1
Specifications
  • Type:
    UV protection
  • Colour:
    Transparent
  • Lenses:
    Clear PC