Orders LinkContactUsLinkComponent
Face shield, polycarbonate
Face shield, polycarbonate
Face shield 5E-11
Catalog # MMMA5E-11
Supplier:   3M
Face shield, polycarbonate
Catalog # MMMA5E-11
Supplier:  3M
Face shield, polycarbonate
Face shield, polycarbonate
Face shield 5E-11
Price
55,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Face Shield 5E-11 is an optical class 1, shade 5, green tinted, curved polycarbonate faceshield with anti-fog and anti-scratch coating for protection against high speed particles, UV, and intense visible light. It is compatible with the V5 Faceshield System for mounting on 3M Hard Hat Series G3000 or G22.
  • Provides protection against high speed particles and secondary welding light
  • Designed to fit 3M Hard Hat Series G3000 or G22 with 3M V5 Faceshield Mounting system
  • Snap-in feature makes it quick and easy to connect to the face shield holder
  • Medium energy impact, Grade B
  • Optical class 1 visor, low energy impact
3M Face Ahield, 5E-11 provides impact protection B against medium speed particles (0,86 g steel ball at 120 m/s), UV, or intense visible light. This anti-fog and anti-scratch coated, shade 5, green tinted, curved polycarbonate faceshield is optical class 1 and Grade B for medium energy impact protection. The curved shape allows for a close fit to the face.
Certifications: EN 166:2001.
Specifications
  • Description:
    Face shield 5E-11
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.