Order Entry
Belgium
ContactUsLinkComponent
Avantor® Seradigm Australia Origin Foetal Bovine Serum (FBS)
Avantor® Seradigm Australia Origin Foetal Bovine Serum (FBS)
Supplier:  VWR Seradigm
Certificates
undefined
Avantor® Seradigm Australia Origin Foetal Bovine Serum (FBS)
Supplier:  VWR Seradigm
Certificates

Specifications

  • Catalog No:
  • 76294-120
  • 76294-122
  • 76294-178
  • 76294-180
  • 76294-124
  • 76294-182
  • Origin:
  • AU
  • AU
  • AU
  • AU
  • AU
  • AU
  • Endotoxin:
  • ≤10 EU/ml
  • ≤10 EU/ml
  • ≤10 EU/ml
  • ≤10 EU/ml
  • ≤10 EU/ml
  • ≤10 EU/ml
  • Haemoglobin:
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • Storage temperature:
  • Frozen
  • Frozen
  • Frozen
  • Frozen
  • Frozen
  • Frozen

Specifications

Related Information

Frequently Bought Together