Orders LinkContactUsLinkComponent
Diffusor for ring light, VisiLED, S40-75
Diffusor for ring light, VisiLED, S40-75
Diffusor for LED ring light, adapting thread M90×1, for S40-75
Catalog # SCOC400580
Diffusor for ring light, VisiLED, S40-75
Catalog # SCOC400580
Diffusor for ring light, VisiLED, S40-75
Diffusor for ring light, VisiLED, S40-75
Diffusor for LED ring light, adapting thread M90×1, for S40-75
Price
319,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The diffusor for LED ring lights avoids reflections of LEDs for uniform illumination.
  • Diffusor with adapting thread (M110×1) to avoid reflections of LEDs
Designed for use only with VisiLED ringlight S40-75.
Ordering information: VisiLED ringlight S40-75 and controller MC 750, MC 1100, or MC 1500 required.
Specifications
  • Description:
    Diffusor for LED ring light, adapting thread M90×1, for S40-75