Order Entry
Belgium
ContactUsLinkComponent
Corning® Mouse native epidermal growth factor (from male submaxillary glands)
Corning® Mouse native epidermal growth factor (from male submaxillary glands)
Catalog # 734-1304
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  
Corning® Mouse native epidermal growth factor (from male submaxillary glands)
Catalog # 734-1304
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Native
  • Source:
    Male submaxillary glands
  • Species:
    Mouse
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Storage conditions:
    Stable for at least three months at 2 to 8 °C (lyophilised) or ≤ three months at −20 °C (solubilised).
    Do not store in frost-free freezer.  Avoid multiple freeze-thaw cycles.
  • Environmental certification:
    ISO 14001 - Bedford
  • Protein/peptide name:
    Epidermal Growth Factor
  • Purity:
    Purity >95% by SDS-PAGE and >80% by pH 9,9 PAGE
    Tested for ability to stimulate proliferation of human foreskin fibroblasts in serum-containing
    medium
    Filtered (0,2 µm membrane) and tested and found negative for bacteria, fungi and mycoplasma
  • Molecular weight:
    6,1 kD
  • Concentration:
    5 - 100 ng/ml medium depending on cell type
  • Formulation:
    Lyophilised from ammonium acetate, carrier-free; reconstitute in dH₂O

Specifications

About this item

Epidermal growth factor (EGF) is a low-molecular weight mitogenic protein that stimulates proliferation of a wide variety of cell types in vitro. EGF can also be used for receptor, gene expression, wound healing studies, and to culture cells in reduced-serum or serum-free culture systems.

EGF can also be used for receptor, gene expression, wound healing studies, and to culture cells in reduced-serum or serum-free culture systems.

Mouse natural EGF are tested for ability to stimulate proliferation of human foreskin fibroblasts in serum-containing medium. Human recombinant EGF are shown to stimulate proliferation of human foreskin fibroblasts in serum-containing medium. All products are filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.