Order Entry
Belgium
ContactUsLinkComponent
Analytical standard, Diofenolan, reference material, Fluka™
Analytical standard, Diofenolan, reference material, Fluka™
  HONCR1722-100MG
 :  
Analytical standard, Diofenolan, reference material, Fluka™
  HONCR1722-100MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Standards:
    Reference material
  • CAS No.:
    63837-33-2
  • Description:
    Diofenolan
  • Description:
    Diofenolan
  • Form:
    Neat