Orders LinkContactUsLinkComponent
Selenium AAS Standards, (CRMs), Fluka™
Supplier:   Honeywell Chemicals
Selenium AAS Standards, (CRMs), Fluka™
Certificates
Selenium AAS Standards, (CRMs), Fluka™
3 Options Available Below
Product Details & Documents
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
 : ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Product Family Options
Product Information
  • Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • Selenium
    Se
    1000 mg/L
    2% HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 34,70 €
    34,70 €
    1 * 100 mL
       34,70 €
  • Selenium
    Se
    1000 mg/L
    2% HNO₃
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 56,70 €
    56,70 €
    1 * 500 mL
       56,70 €
  • Selenium
    Se
    10000 mg/L
    5% HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 139,00 €
    139,00 €
    1 * 100 mL
       139,00 €