Order Entry
Belgium
ContactUsLinkComponent
Mercury ICP and ICP-MS standards, (CRMs), Fluka™
Mercury ICP and ICP-MS standards, (CRMs), Fluka™
Catalog # HONCE1391-100ML
CAS Number:  
Mercury ICP and ICP-MS standards, (CRMs), Fluka™
Catalog # HONCE1391-100ML
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Mercury
  • Symbol:
    Hg

Specifications

About this item

Certified Reference Materials (CRMs)

  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).