Order Entry
Belgium
Orders LinkContactUsLinkComponent
Selenium ICP Standards, (CRMs), Fluka™

Selenium ICP Standards, (CRMs), Fluka™

Supplier:   Honeywell Chemicals
Selenium ICP Standards, (CRMs), Fluka™
Certificates
Selenium ICP Standards, (CRMs), Fluka™

2 Options Available Below

Product Details & Documents

Certified Reference Materials (CRMs)

  • Traceable to SRM from NIST

Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).

Product Family Options

Product Information

  • Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • Selenium
    Se
    1000 mg/L
    2% HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 79,20 €
    79,20 €
    1 * 100 mL
       79,20 €

  • Selenium
    Se
    10000 mg/L
    5% HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 152,00 €
    152,00 €
    1 * 100 mL
       152,00 €