ContactUsLinkComponent
Upright laboratory microscopes, DM2000 LED
Upright laboratory microscopes, DM2000 LED
Supplier:  LEICA MICROSYSTEMS
Certificates
undefined
Upright laboratory microscopes, DM2000 LED
Supplier:  LEICA MICROSYSTEMS
Specifications
  • Catalog No:
  • 630-3184
  • 630-3182
  • 630-3183
  • 630-3185
  • Model:
  • DM 2000 (Package 3), PH
  • DM 2000 (Package 1), BF
  • DM 2000 (Package 2), BF
  • DM 2000 (Package 4), PH
  • Type:
  • Binocular
  • Binocular
  • Trinocular
  • Trinocular
  • Observation method:
  • Phase contrast
  • Brightfield
  • Brightfield
  • Phase contrast
  • Eyepieces:
  • WF 10×/22 mm
  • WF 10×/22 mm
  • WF 10×/22 mm
  • WF 10×/22 mm
  • Nosepiece:
  • Sextuple
  • Sextuple
  • Sextuple
  • Sextuple
  • Objectives:
  • 4×, 10×, 20×, 40×, 100× (i)
  • 4×, 10×, 20×, 40×, 100× (i)
  • 4×, 10×, 20×, 40×, 100× (i)
  • 4×, 10×, 20×, 40×, 100× (i)
  • Illumination:
  • LED
  • LED
  • LED
  • LED