Order Entry
Belgium
Orders LinkContactUsLinkComponent
Multi-element standard solution 4 for ICP

Multi-element standard solution 4 for ICP

Multielement Standard Solution 4, 10% HNO₃, Plastic bottle, 100 mL
Catalog # HONCE2233-100ML
Supplier:   Honeywell Chemicals
Multi-element standard solution 4 for ICP
Catalog # HONCE2233-100ML
CAS Number:  
Multi-element standard solution 4 for ICP

Multielement Standard Solution 4, 10% HNO₃, Plastic bottle, 100 mL

CAS Number:  
Price
197,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Certified Reference Material (CRM).

100 mg/L each of B, Fe, Se, Tl, Zn, 40 mg/L each of Al, As, Ba, Pb, V, 20 mg/L each of Cr, Cu, Ni, 10 mg/L each of Be, Cd, Co, Mn in HNO₃ 10%

Specifications

Specifications

  • Matrix:
    10% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multielement Standard Solution 4

Specifications