Order Entry
Belgium
ContactUsLinkComponent
 
Multi-element standard solution 3 for ICP
Catalog # HONCE2231-100ML
CAS Number:  
Multi-element standard solution 3 for ICP
Catalog # HONCE2231-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multielement Standard Solution 3

Specifications

About this item

Certified Reference Material (CRM).

2000 mg/L Ca, 1000 mg/L Na, 400 mg/L Mg, 200 mg/L K in HNO₃ 5%