Order Entry
Belgium
ContactUsLinkComponent
Multi-element standard solution 1 for ICP
Multi-element standard solution 1 for ICP
Catalog # HONCE2230-100ML
CAS Number:  
Multi-element standard solution 1 for ICP
Catalog # HONCE2230-100ML
CAS Number:  

Specifications

  • Matrix:
    10% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multielement Standard Solution 1

Specifications

About this item

Certified Reference Material (CRM).

100 mg/L each of Bi, K, Pb, 50 mg/L each of Al, B, Cr, Li, Mo, Na, Ni, Tl, 10 mg/L each of Ag, Ba, Ca, Cd, Co, Cu, Fe, Mg, Mn, Sr, Zn in HNO₃ 10%