Order Entry
Belgium
ContactUsLinkComponent
Concentrated Eluent for Ion Chromatography, Methanesulfonic Acid Solution
Concentrated Eluent for Ion Chromatography, Methanesulfonic Acid Solution
Catalog # HONCE2211-500ML
CAS Number:  
Concentrated Eluent for Ion Chromatography, Methanesulfonic Acid Solution
Catalog # HONCE2211-500ML
CAS Number:  

Specifications

  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Concentrated Eluent for Ion Chromatography, Methanesulfonic Acid Solution

Specifications

About this item

Certified Reference Material (CRM).

0.5 M Methanesulfonic acid in water