Order Entry
Belgium
ContactUsLinkComponent
 
Concentrated Eluent for Ion Chromatography, Sodium Carbonate/Sodium Bicarbonate Solution 3
Catalog # HONCE2209-500ML
CAS Number:  
Concentrated Eluent for Ion Chromatography, Sodium Carbonate/Sodium Bicarbonate Solution 3
Catalog # HONCE2209-500ML
CAS Number:  

Specifications

  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Concentrated Eluent for Ion Chromatography, Sodium Carbonate/Sodium Bicarbonate Solution 3

Specifications

About this item

Certified Reference Material (CRM).

0.35 M Na₂CO₃ / 0.10 M NaHCO₃ in water