Concentrated Eluent for Ion Chromatography, Sodium Carbonate/Sodium Bicarbonate Solution 1
Catalog # HONCE2207-500ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:500 mL
- Description:Concentrated Eluent for Ion Chromatography, Sodium Carbonate/Sodium Bicarbonate Solution 1
Specifications
About this item
Certified Reference Material (CRM).
0.022M Na₂CO₃ / 0.28M NaHCO₃ in water