Order Entry
Belgium
ContactUsLinkComponent
GFAAS Mixed Standard
GFAAS Mixed Standard
Catalog # HONCE2203-100ML
CAS Number:  
GFAAS Mixed Standard
Catalog # HONCE2203-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃/tr HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    GFAAS Mixed Standard

Specifications

About this item

Certified Reference Material (CRM).

100 mg/L each of Al, As, Pb, Sb, Se, Tl, 50 mg/L each of Ba, Co, Cu, Ni, 20 mg/L each of Cr, Fe, Mn, 10 mg/L Ag, 5 mg/L each of Be, Cd in HNO₃ 5%, tr. HF