Order Entry
Belgium
Orders LinkContactUsLinkComponent
NexION AFT single-element solution

NexION AFT single-element solution

NexION AFT Single-Element Solution, 2% HNO₃, Plastic bottle, 100 mL
Catalog # HONCE2196-100ML
Supplier:   Honeywell Chemicals
NexION AFT single-element solution
Catalog # HONCE2196-100ML
CAS Number:  
NexION AFT single-element solution

NexION AFT Single-Element Solution, 2% HNO₃, Plastic bottle, 100 mL

CAS Number:  
Price
123,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Certified Reference Material (CRM).

2 g/L Iron (Fe) in HNO₃ 2%

Specifications

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    NexION AFT Single-Element Solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.