Order Entry
Belgium
ContactUsLinkComponent
NexION standard DRC mode detection limit solution
NexION standard DRC mode detection limit solution
Catalog # HONCE2193-100ML
CAS Number:  
NexION standard DRC mode detection limit solution
Catalog # HONCE2193-100ML
CAS Number:  

Specifications

  • Matrix:
    0,5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    NexION Standard DRC Mode Detection Limit Solution

Specifications

About this item

Certified Reference Material (CRM).

1 g/L each of Be, Ca, Co, Fe, In, U in HNO₃ 0.5%