Order Entry
Belgium
ContactUsLinkComponent
NexION setup solution
NexION setup solution
Catalog # HONCE2188-100ML
CAS Number:  
NexION setup solution
Catalog # HONCE2188-100ML
CAS Number:  

Specifications

  • Matrix:
    1% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    NexION Setup Solution

Specifications

About this item

Certified Reference Material (CRM).

1 g/L each of Be, Ce, Fe, In, Li, Mg, Pb, U in HNO₃ 1%