Order Entry
Belgium
ContactUsLinkComponent
SmartTune solution for DRC/DRCplus/DRC II Hg for ICP
SmartTune solution for DRC/DRCplus/DRC II Hg for ICP
Catalog # HONCE2187-500ML
CAS Number:  
SmartTune solution for DRC/DRCplus/DRC II Hg for ICP
Catalog # HONCE2187-500ML
CAS Number:  

Specifications

  • Matrix:
    0,5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    SmartTune Solution

Specifications

About this item

Certified Reference Material (CRM).

10 g/L Ba; 1 g/L each of Be, Ce, Co, In, Fe, Pb, Mg, Th, U in HNO₃ 0.5%