Order Entry
Belgium
ContactUsLinkComponent
ELAN 6100 DRC setup stab masscal solution Hg for ICP
ELAN 6100 DRC setup stab masscal solution Hg for ICP
Catalog # HONCE2182-100ML
CAS Number:  
ELAN 6100 DRC setup stab masscal solution Hg for ICP
Catalog # HONCE2182-100ML
CAS Number:  

Specifications

  • Matrix:
    0,5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    ELAN 6100 DRC Setup Stab Masscal Solution Hg

Specifications

About this item

Certified Reference Material (CRM).

10 g/L Ba; 1 g/L each of Al, Cd, Ce, Cr, Cu, In, Pb, Mg, Mn, Rh, Th in HNO₃ 0.5%