Order Entry
Belgium
ContactUsLinkComponent
Multi-element solution 1 for AA and ICP-OES
Multi-element solution 1 for AA and ICP-OES
Catalog # HONCE2177-100ML
CAS Number:  
Multi-element solution 1 for AA and ICP-OES
Catalog # HONCE2177-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-Element Solution 1

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L each of Be, Bi, Ce, Co, In, Mg, Ni, Pb, U in HNO₃ 2%