Order Entry
Belgium
ContactUsLinkComponent
 
Multi-element internal standard for AA and ICP-OES
Catalog # HONCE2175-100ML
CAS Number:  
Multi-element internal standard for AA and ICP-OES
Catalog # HONCE2175-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-Element Internal Standard

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L each of Bi, Ho, In, 6Li, Sc, Tb, Y in HNO₃ 2%