Multi-element solution 4 for AA and ICP-OES
Multi-Element Solution 4, 10% HCl/1% HNO₃, Plastic bottle, 100 mL
Catalog # HONCE2174-100ML
Supplier: Honeywell Chemicals
Multi-element solution 4 for AA and ICP-OES
Catalog # HONCE2174-100ML
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Certified Reference Material (CRM).
10 mg/L each of Au, Hf, Ir, Pd, Pt, Rh, Ru, Sb, Sn, Te in HCl 10%, HNO₃ 1%
Specifications
Specifications
- Matrix:10% HCl/1% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Description:Multi-Element Solution 4
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



