Order Entry
Belgium
ContactUsLinkComponent
Universal Data Acquisition Standard 1 for ICP
Universal Data Acquisition Standard 1 for ICP
Catalog # HONCE2221-100ML
CAS Number:  
Universal Data Acquisition Standard 1 for ICP
Catalog # HONCE2221-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Universal Data Acquisition Standard 1

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L each of Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sm, Sc, Tb, Th, Tm, Y, Yb in HNO₃ 5%