Order Entry
Belgium
ContactUsLinkComponent
internal standard mix for ICP
internal standard mix for ICP
Catalog # HONCE2172-100ML
CAS Number:  
internal standard mix for ICP
Catalog # HONCE2172-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Internal Standard Mix

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L each of 6Li, Sc, Ge, Y, In, Tb, Bi in HNO₃ 5%