Order Entry
Belgium
ContactUsLinkComponent
Alternate Metals I for AA and ICP-OES
Alternate Metals I for AA and ICP-OES
Catalog # HONCE2169-100ML
CAS Number:  
Alternate Metals I for AA and ICP-OES
Catalog # HONCE2169-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Alternate Metals I

Specifications

About this item

Certified Reference Material (CRM).

20 mg/L each of Al, Fe, V, 10 mg/L each of Co, Cu, Mn, Ni, Zn, 5 mg/L each of Be, Sb, Tl in HNO₃ 2%