Order Entry
Belgium
ContactUsLinkComponent
Trace Metals II for AA and ICP-OES
Trace Metals II for AA and ICP-OES
Catalog # HONCE2167-100ML
CAS Number:  
Trace Metals II for AA and ICP-OES
Catalog # HONCE2167-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Trace Metals II

Specifications

About this item

Certified Reference Material (CRM).

20 mg/L each of Sb, Tl, 10 mg/L Ag in HNO₃ 2%