Order Entry
Belgium
Orders LinkContactUsLinkComponent
USP 232 elemental impurities solution 2

USP 232 elemental impurities solution 2

USP 232 Elemental Impurities Solution 2, 15% HCl, Plastic bottle, 100 mL
Catalog # HONCE2163-100ML
Supplier:   Honeywell Chemicals
USP 232 elemental impurities solution 2
Catalog # HONCE2163-100ML
CAS Number:  
USP 232 elemental impurities solution 2

USP 232 Elemental Impurities Solution 2, 15% HCl, Plastic bottle, 100 mL

CAS Number:  
Price
521,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Certified Reference Material (CRM).

100 mg/kg each of Ir, Os, Pd, Pt, Rh, Ru in HCl 15%

Specifications

Specifications

  • Matrix:
    15% HCl
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    USP 232 Elemental Impurities Solution 2

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.