Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Interference A for ICP

Interference A, 5% HNO₃/tr HF, Plastic bottle, 100 mL
Catalog # HONCE2155-100ML
Supplier:   Honeywell Chemicals
Interference A for ICP
Catalog # HONCE2155-100ML
CAS Number:  
Interference A for ICP

Interference A, 5% HNO₃/tr HF, Plastic bottle, 100 mL

CAS Number:  
Price
548,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Certified Reference Material (CRM).

21215 mg/L Cl-, 3000 mg/L Ca, 2500 mg/L each of Fe, Na, 2000 mg/L C, 1000 mg/L each of Al, K, Mg, P, S, 20 mg/L each of Mo, Ti in Nitric Acid 5% in HNO₃ 5%

Specifications

Specifications

  • Matrix:
    5% HNO₃/tr HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Interference A

Specifications