Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Multi-element solution for ICP, (3 components)

Multi-Element Solution for ICP, (3 components), 5% HNO₃, Plastic bottle, 100 mL
Catalog # HONCE2132-100ML
Supplier:   Honeywell Chemicals
Multi-element solution for ICP, (3 components)
Catalog # HONCE2132-100ML
CAS Number:  
Multi-element solution for ICP, (3 components)

Multi-Element Solution for ICP, (3 components), 5% HNO₃, Plastic bottle, 100 mL

CAS Number:  
Price
311,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Certified Reference Material (CRM).

1000 mg/L each of K, Na, P in HNO₃ 5%

Specifications

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-Element Solution for ICP, (3 components)

Specifications