Order Entry
Belgium
ContactUsLinkComponent
Spike sample standard soil for ICP
Spike sample standard soil for ICP
Catalog # HONCE2129-100ML
CAS Number:  
Spike sample standard soil for ICP
Catalog # HONCE2129-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Spike Sample Standard Soil

Specifications

About this item

Certified Reference Material (CRM).

250 mg/L each of Ba, Cr, Cu, Zn, 150 mg/L V, 125 mg/L Ni, 100 mg/L each of Co, Pb, Sb, 50 mg/L each of As, Cd, 25 mg/L each of Ag, Be, Se, Tl in HNO₃ 5%