ContactUsLinkComponent
Spike sample standard I for ICP
Spike sample standard I for ICP
  HONCE2127-100ML
 :  
Spike sample standard I for ICP
  HONCE2127-100ML
 :  
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Spike Sample Standard I
Certified Reference Material (CRM).
200 mg/L each of Al, As, Ba, Se, Tl, 100 mg/L Fe, 50 mg/L each of Co, Mn, Ni, Pb, Sb, V, Zn, 25 mg/L Cu, 20 mg/L Cr, 5 mg/L each of Ag, Be, Cd in HNO₃ 5%