Order Entry
Belgium
Orders LinkContactUsLinkComponent
VIS wavecal solution for ICP

VIS wavecal solution for ICP

VIS Wavecal Solution, 2% HNO₃, Plastic bottle, 100 mL
Catalog # HONCE2122-100ML
Supplier:   Honeywell Chemicals
VIS wavecal solution for ICP
Catalog # HONCE2122-100ML
CAS Number:  
VIS wavecal solution for ICP

VIS Wavecal Solution, 2% HNO₃, Plastic bottle, 100 mL

CAS Number:  
Price
195,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Certified Reference Material (CRM).

50 mg/L K, 10 mg/L each of La, Li, Mn, Na, Sr, 1 mg/L each of Ba, Ca in HNO₃ 2%

Specifications

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    VIS Wavecal Solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.